Merck
  • Palladium-catalyzed cyanomethylation of aryl halides through domino Suzuki coupling-isoxazole fragmentation.
Palladium-catalyzed cyanomethylation of aryl halides through domino Suzuki coupling-isoxazole fragmentation.
Journal of the American Chemical Society (2011-04-21)
Juraj Velcicky, Arne Soicke, Roland Steiner, Hans-Günther Schmalz
ABSTRACT
A one-pot protocol for the cyanomethylation of aryl halides through a palladium-catalyzed reaction with isoxazole-4-boronic acid pinacol ester was developed. Mechanistically, the reaction proceeds through (1) Suzuki coupling, (2) base-induced fragmentation, and (3) deformylation as shown by characterization of all postulated intermediates. Under optimized conditions (PdCl(2)dppf, KF, DMSO/H(2)O, 130 °C) a broad spectrum of aryl bromides could be converted into arylacetonitriles with up to 88% yield.
Sigma-Aldrich
Palladium(II) chloride, ReagentPlus®, 99%
Sigma-Aldrich
Palladium(II) chloride, anhydrous, 59-60% palladium (Pd) basis
Sigma-Aldrich
Palladium(II) chloride, 99.995%
Sigma-Aldrich
Palladium(II) chloride, anhydrous, 60% Pd basis
Sigma-Aldrich
Palladium(II) chloride, ≥99.9%
Sigma-Aldrich
Palladium(II) chloride solution, 5 wt. % in 10 wt. % HCl
Sigma-Aldrich
Palladium(II) chloride ChemBeads