Skip to Content
Merck
All Photos(1)

Documents

270245

Sigma-Aldrich

4-Methoxybenzyl chloride

contains potassium carbonate as stabilizer, 98%

Synonym(s):

4-(Chloromethyl)anisole

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H4CH2Cl
CAS Number:
Molecular Weight:
156.61
Beilstein:
606667
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

contains

potassium carbonate as stabilizer

refractive index

n20/D 1.548 (lit.)

bp

117-118 °C/14 mmHg (lit.)

mp

−1 °C (lit.)

density

1.155 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

COc1ccc(CCl)cc1

InChI

1S/C8H9ClO/c1-10-8-4-2-7(6-9)3-5-8/h2-5H,6H2,1H3

InChI key

MOHYOXXOKFQHDC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Methoxybenzyl chloride undergoes mild oxidative claevage with ceric ammonium nitrate.

Application

4-Methoxybenzyl chloride was used to benzylate the aniline nitrogen. It was also used in the synthesis of 1-naphthamide and diarymethanes via Suzuki cross-coupling with postassium aryltrifluoroborates.
O/N protecting group reagent which can be mildly oxidatively cleaved, e.g., with ceric ammonium nitrate.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

228.2 °F - closed cup

Flash Point(C)

109 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gary A Molander et al.
The Journal of organic chemistry, 71(24), 9198-9202 (2006-11-18)
[reaction: see text] The palladium-catalyzed cross-coupling of potassium aryltrifluoroborates with benzylic halides occurs in good yield with high functional group tolerance. The increased stability of potassium aryltrifluoroborates compared to other boron coupling partners makes this an effective route to functionalized
Martin H Bolli et al.
Journal of medicinal chemistry, 47(11), 2776-2795 (2004-05-14)
Since its discovery in 1988 by Yanagisawa et al., endothelin (ET), a potent vasoconstrictor, has been widely implicated in the pathophysiology of cardiovascular, cerebrovascular, and renal diseases. Many research groups have embarked on the discovery and development of ET receptor
Andrew J Horan et al.
Analytical chemistry, 89(2), 1059-1062 (2017-02-15)
Airborne nanoparticles play a key role in climate effects as well as impacting human health. Their small mass and complex chemical composition represent significant challenges for analysis. This work introduces a new ionization method, droplet assisted inlet ionization (DAII), where
Thompson AS, et al.
Tetrahedron Letters, 36(49), 8937-8940 (1995)
Synthesis of a potent (?)-4-(2-hydroxyphenyl) analogue of the acromelic acids by dearomatising cyclisation of a lithiated N-p-methoxybenzyl-4-methoxy-1-naphthamide.
Tetrahedron Letters, 42(20), 3407-3410 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service