![4-Methoxybenzyl chloride](/deepweb/assets/sigmaaldrich/product/structures/797/367/ebe34d22-5bc1-4e72-9bd2-37ece98938e5/640/ebe34d22-5bc1-4e72-9bd2-37ece98938e5.png)
4-Methoxybenzyl chloride
![4-Methoxybenzyl chloride](/deepweb/assets/sigmaaldrich/product/structures/797/367/ebe34d22-5bc1-4e72-9bd2-37ece98938e5/640/ebe34d22-5bc1-4e72-9bd2-37ece98938e5.png)
Synonym(s):
4-(Chloromethyl)anisole
Linear Formula:
CH3OC6H4CH2Cl
CAS Number:
Molecular Weight:
156.61
Products
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing