Skip to Content
Merck
All Photos(1)

Key Documents

B83606

Sigma-Aldrich

4-Methylbenzyl bromide

97%

Synonym(s):

p-Xylyl bromide

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€64.30

€64.30


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
€64.30

About This Item

Linear Formula:
CH3C6H4CH2Br
CAS Number:
Molecular Weight:
185.06
Beilstein:
507389
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€64.30


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

powder

impurities

<1.5% 1,4-bis(bromomethyl)benzene

bp

218-220 °C (lit.)

mp

34-36 °C (lit.)

storage temp.

2-8°C

SMILES string

Cc1ccc(CBr)cc1

InChI

1S/C8H9Br/c1-7-2-4-8(6-9)5-3-7/h2-5H,6H2,1H3

InChI key

WZRKSPFYXUXINF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

208.4 °F - closed cup

Flash Point(C)

98 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Zhouyang Li et al.
Talanta, 174, 198-205 (2017-07-26)
This paper reports novel imidazole fluorescent poly(ionic liquid) nanoparticles (FPILNs) of poly(1-[(4-methyphenyl)methyl]-3-vinyl-imidazolium bromide (poly([MVI]Br) for selective and sensitive determination of pyrogallol. An imidazole ionic liquid of 1-[(4-methyphenyl)methyl]-3-vinyl-imidazolium bromide ([MVI]Br) was synthesized and used as the only monomer to obtain poly([MVI]Br)
Ngoc-Duc Doan et al.
Journal of peptide science : an official publication of the European Peptide Society, 21(5), 387-391 (2014-11-18)
The solid-phase synthesis of azapeptides possessing a C-terminal aza-residue has been accomplished by a protocol featuring regioselective alkylation of benzhydrylidene-aza-glycinamide and illustrated by the syntheses of [aza-Lys(6)] growth-hormone-releasing peptide-6 analogs.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service