Skip to Content
Merck
All Photos(2)

Key Documents

263931

Sigma-Aldrich

Ethyl-tetramethylcyclopentadiene

97%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€340.00

€340.00


Estimated to ship on13 April 2025


Request a Bulk Order

Select a Size

Change View
5 G
€340.00

About This Item

Empirical Formula (Hill Notation):
C11H18
CAS Number:
Molecular Weight:
150.26
Beilstein:
2349373
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€340.00


Estimated to ship on13 April 2025


Request a Bulk Order

Assay

97%

refractive index

n20/D 1.479 (lit.)

bp

60-62 °C/5 mmHg (lit.)

density

0.841 g/mL at 25 °C (lit.)

SMILES string

CCC1C(C)=C(C)C(C)=C1C

InChI

1S/C11H18/c1-6-11-9(4)7(2)8(3)10(11)5/h11H,6H2,1-5H3

InChI key

MXYDFFLUOPTQFI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Ethyl-tetramethylcyclopentadiene may be used in chemical synthesis.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

131.0 °F - closed cup

Flash Point(C)

55 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service