Skip to Content
Merck
All Photos(1)

Documents

520276

Sigma-Aldrich

Diethyldicyclopentadiene,mixture of isomers

Synonym(s):

Diethyl-3a,4,7,7a-tetrahydro-4,7-methano-1H-indene, Ethylcyclopentadiene dimer

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H20
CAS Number:
Molecular Weight:
188.31
UNSPSC Code:
12352100
NACRES:
NA.22

refractive index

n20/D 1.496 (lit.)

bp

191 °C (lit.)

density

0.924 g/mL at 25 °C (lit.)

InChI

1S/C14H20/c1-3-14(4-2)8-7-12-10-5-6-11(9-10)13(12)14/h5-8,10-13H,3-4,9H2,1-2H3

InChI key

QWQMAINGVKHIQJ-UHFFFAOYSA-N

Related Categories

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

185.0 °F - closed cup

Flash Point(C)

85 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service