Skip to Content
Merck
All Photos(1)

Key Documents

8.18534

Sigma-Aldrich

1,2,3,4,5-Pentamethylcyclopentadiene

for synthesis

Synonym(s):

1,2,3,4,5-Pentamethylcyclopentadiene

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
€45.80
5 ML
€163.00

€45.80


0 left in stock

Request a Bulk Order

Select a Size

Change View
1 ML
€45.80
5 ML
€163.00

About This Item

Empirical Formula (Hill Notation):
C10H16
CAS Number:
Molecular Weight:
136.23
MDL number:
UNSPSC Code:
12352002
EC Index Number:
223-743-4
NACRES:
NA.22

€45.80


0 left in stock

Request a Bulk Order

Quality Level

Assay

≥88.0% (GC)

form

liquid

bp

58 °C/17 hPa

transition temp

flash point 44 °C

density

0.84 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

C1(C(=C(C(=C1C)C)C)C)C

InChI

1S/C10H16/c1-6-7(2)9(4)10(5)8(6)3/h6H,1-5H3

InChI key

WQIQNKQYEUMPBM-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 88.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.837 - 0.843
Identity (IR): passes test

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

111.2 °F - closed cup

Flash Point(C)

44 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service