Skip to Content
Merck
All Photos(1)

Key Documents

261653

Sigma-Aldrich

Benzoyl isothiocyanate

98%

Synonym(s):

Benzoylthiocarbimide, N-Benzoyl isothiocyanate

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€60.70
25 G
€105.00

€60.70


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€60.70
25 G
€105.00

About This Item

Linear Formula:
C6H5CONCS
CAS Number:
Molecular Weight:
163.20
Beilstein:
606716
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€60.70


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.6354 (lit.)

bp

128-131 °C/15 mmHg (lit.)

density

1.214 g/mL at 25 °C (lit.)

functional group

amine
isothiocyanate
phenyl

storage temp.

2-8°C

SMILES string

O=C(N=C=S)c1ccccc1

InChI

1S/C8H5NOS/c10-8(9-6-11)7-4-2-1-3-5-7/h1-5H

InChI key

CPEKAXYCDKETEN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Benzoyl isothiocyanate has been used in the preparation of:
  • 1-benzoyl-3-(5-chloro-2-hydroxyphenyl)thiourea, new thiourea derivative[1]
  • guanine[2]

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Skin Sens. 1

Supplementary Hazards

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Madhusudhana Reddy et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 186, 8-16 (2017-06-11)
Sensing of F
Synthesis of guanosine and its derivatives from 5-amino-1-beta-d-ribofuranoxyl-4-imidazolecarboxamide. I. Ring closure with benzoyl isothiocyanate.
A Yamazaki et al.
The Journal of organic chemistry, 32(6), 1825-1828 (1967-06-01)
Murat Atiş et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 98, 290-301 (2012-09-25)
A new thiourea derivative, 1-benzoyl-3-(5-chloro-2-hydroxyphenyl)thiourea (bcht) has been synthesized from the reaction of 2-amino-4-chlorophenol with benzoyl isothiocyanate. The title compound has been characterized by elemental analyses, FT-IR, (13)C, (1)H NMR spectroscopy and the single crystal X-ray diffraction analysis. The structure

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service