Skip to Content
Merck
All Photos(1)

Key Documents

CDS020502

Sigma-Aldrich

2-Aminoimidazole

AldrichCPR

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€59.60

€59.60


Ships within 2 business days(Orders outside of US, please allow an additional 1-2 weeks for delivery)

Request a Bulk Order

Select a Size

Change View
10 MG
€59.60

About This Item

Empirical Formula (Hill Notation):
C3H5N3
CAS Number:
Molecular Weight:
83.09
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

€59.60


Ships within 2 business days(Orders outside of US, please allow an additional 1-2 weeks for delivery)

Request a Bulk Order

form

crystals

SMILES string

Nc1ncc[nH]1

InChI

1S/C3H5N3/c4-3-5-1-2-6-3/h1-2H,(H3,4,5,6)

InChI key

DEPDDPLQZYCHOH-UHFFFAOYSA-N

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marina Gjorgjieva et al.
Mini reviews in medicinal chemistry, 18(19), 1640-1658 (2016-05-06)
Pyrrole-2-aminoimidazole compounds are found in marine organisms, mainly as secondary metabolites in various marine sponges. Studies of natural pyrrole-2-aminoimidazole compounds showed that they possess different pharmacological properties, such as antimicrobial, antibiofilm, immunosuppressive and anticancer activities. Many analogs of the natural
Travis Walton et al.
Biochemistry, 56(43), 5739-5747 (2017-10-13)
The nonenzymatic polymerization of RNA may have enabled copying of functional sequences during the origin of life. Recent progress utilizing 5'-phosphoro-2-aminoimidazole activation has reinvigorated the possibility of using nonenzymatic RNA polymerization for copying arbitrary sequences. However, the reasons why 2-aminoimidazole
Michael K Fenwick et al.
Proceedings of the National Academy of Sciences of the United States of America, 114(26), 6806-6811 (2017-06-14)
Viperin is an IFN-inducible radical
Amrita B Hazra et al.
Proceedings of the National Academy of Sciences of the United States of America, 112(34), 10792-10797 (2015-08-08)
Vitamin B12 (cobalamin) is required by humans and other organisms for diverse metabolic processes, although only a subset of prokaryotes is capable of synthesizing B12 and other cobamide cofactors. The complete aerobic and anaerobic pathways for the de novo biosynthesis
Ulrike Klenke et al.
Endocrinology, 155(5), 1851-1863 (2014-02-26)
Metabolic dysfunctions are often linked to reproductive abnormalities. Adiponectin (ADP), a peripheral hormone secreted by white adipose tissue, is important in energy homeostasis and appetite regulation. GnRH neurons are integral components of the reproductive axis, controlling synthesis, and release of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service