Skip to Content
Merck
All Photos(1)

Documents

259462

Sigma-Aldrich

Diethoxy(methyl)vinylsilane

97%

Synonym(s):

(Diethoxymethylsilyl)ethylene, Methylvinyldiethoxysilane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3Si(OC2H5)2CH=CH2
CAS Number:
Molecular Weight:
160.29
Beilstein:
970124
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

Assay

97%

form

liquid

refractive index

n20/D 1.401 (lit.)

bp

133-134 °C (lit.)

density

0.858 g/mL at 25 °C (lit.)

SMILES string

CCO[Si](C)(OCC)C=C

InChI

1S/C7H16O2Si/c1-5-8-10(4,7-3)9-6-2/h7H,3,5-6H2,1-2,4H3

InChI key

MBGQQKKTDDNCSG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Diethoxy(methyl)vinylsilane is an organosiloxane with a reactive vinyl group. It has a good toughness which is highly thermo-stable and a good film former. It can be converted into a highly cross-linked polymer through thermal polymerization reaction.

Application

Diethoxy(methyl)vinylsilane can be used to prepare benzocyclobutene(BCB) functionalized siloxanes for the development of encapsulating resins in microelectronics industry. It may be used in the preparation of polysiloxane modified tetraphenylethene(PTPESi) which finds potential application as a sensor for vapour phase detection of explosives. It may also be used in the surface modification of silica matrix that enables better chromatographic properties in cation-exchange process.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

62.6 °F - closed cup

Flash Point(C)

17 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

High performance low dielectric polysiloxanes with high thermostability and low water uptake.
Chen X, et al.
Materials Chemistry Frontiers., 2(7), 1397-1402 (2018)
Synthesis, characterization, and cure kinetics analysis of high refractive index copolysiloxanes.
Zhan X, et al.
Journal of Thermal Analysis and Calorimetry, 117(2), 875-883 (2014)
Polysiloxane-Modified Tetraphenylethene: Synthesis, AIE Properties, and Sensor for Detecting Explosives.
Li Q, et al.
Macromolecular Rapid Communications, 37(21), 1772-1779 (2016)
Maleic acid-styrene encapsulated silica cation exchanger in high performance liquid chromatography.
Yang R, et al.
Talanta, 55(6), 1091-1096 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service