Skip to Content
Merck
All Photos(1)

Key Documents

251194

Sigma-Aldrich

1-Fluoropentane

98%

Synonym(s):

Amyl fluoride

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€147.00

€147.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€147.00

About This Item

Linear Formula:
CH3(CH2)4F
CAS Number:
Molecular Weight:
90.14
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€147.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.36 (lit.)

bp

62-63 °C (lit.)

density

0.789 g/mL at 25 °C (lit.)

functional group

alkyl halide
fluoro

SMILES string

CCCCCF

InChI

1S/C5H11F/c1-2-3-4-5-6/h2-5H2,1H3

InChI key

OEPRBXUJOQLYID-UHFFFAOYSA-N

General description

Dual luminescence of 4-N,N-dimethylaminobenzonitrile has been observed in 1-fluoropentane[1].

Application

1-Fluoropentane has been used in the synthesis of 6-F-B10H13 and 6-Cl- B10H13 (halodecaboranes)[2].

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-9.4 °F - closed cup

Flash Point(C)

-23 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

C F Nhachi et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 26(8), 705-713 (1988-08-01)
Phenobarbitone pretreatment potentiated hepatocyte lesions in male rats 24 hr after treatment with 1-fluoropentane (3.5 mg/kg body weight) and 1-fluorohexane (0.17 mg/kg body weight). Serum levels of the enzymes ornithine carbamyltransferase, glutamic-pyruvic transaminase and gamma-glutamyltranspeptidase were significantly elevated by the
The role of the solvent in the dual luminescence of 4-N, N-dimethylaminobenzonitrile.
Suppan P.
Chemical Physics Letters, 128(2), 160-161 (1986)
K R Harikumar et al.
Nature chemistry, 1(9), 716-721 (2010-12-03)
The controlled imprinting of surfaces with specified patterns is important in the development of nanoscale devices. Previously, such patterns were created using self-assembled physisorbed adsorbate molecules that can be stabilized on the surface by subsequent chemical bonding. Here we show
William C Ewing et al.
Inorganic chemistry, 47(19), 8580-8582 (2008-08-30)
The high-yield syntheses of 6-X-B 10H 13 [X = Cl (88%), Br (96%), I (84%)] resulted from the cage-opening reactions of the (NH 4 (+)) 2B 10H 10 (2-) salt with ionic-liquid-based superacidic hydrogen halides, while both the previously unknown

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service