Skip to Content
Merck
All Photos(1)

Key Documents

250082

Sigma-Aldrich

1-Fluoroheptane

98%

Synonym(s):

Heptyl fluoride

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€61.10

€61.10


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€61.10

About This Item

Linear Formula:
CH3(CH2)6F
CAS Number:
Molecular Weight:
118.19
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€61.10


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.386 (lit.)

bp

119.2 °C/750 mmHg (lit.)

density

0.806 g/mL at 25 °C (lit.)

functional group

alkyl halide
fluoro

SMILES string

CCCCCCCF

InChI

1S/C7H15F/c1-2-3-4-5-6-7-8/h2-7H2,1H3

InChI key

BITLXSQYFZTQGC-UHFFFAOYSA-N

General description

Specific rotations of chiral conformers of 1-fluoroheptane were computed using an approach in which localization is enforced throughout the orbital optimization and subsequent linear response computation[1].

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

57.2 °F - closed cup

Flash Point(C)

14 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Harley R McAlexander et al.
Physical chemistry chemical physics : PCCP, 14(21), 7830-7836 (2012-05-01)
The impact of orbital localization on the efficiency and accuracy of the optimized-orbital coupled cluster model is examined for the prediction of chiroptical properties, in particular optical rotation. The specific rotations of several test cases-(P)-[4]triangulane, (S)-1-phenylethanol, and chiral conformers of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service