Skip to Content
Merck
All Photos(1)

Key Documents

F6001

Sigma-Aldrich

Fluorobenzene

99%

Synonym(s):

Monofluorobenzene, Phenyl fluoride

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€34.40
100 G
€42.40
500 G
€153.00

€34.40


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€34.40
100 G
€42.40
500 G
€153.00

About This Item

Empirical Formula (Hill Notation):
C6H5F
CAS Number:
Molecular Weight:
96.10
Beilstein:
1236623
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€34.40


Check Cart for Availability

Request a Bulk Order

vapor density

3.31 (vs air)

Quality Level

Assay

99%

refractive index

n20/D 1.465 (lit.)

bp

85 °C (lit.)

mp

−42 °C (lit.)

density

1.024 g/mL at 25 °C (lit.)

SMILES string

Fc1ccccc1

InChI

1S/C6H5F/c7-6-4-2-1-3-5-6/h1-5H

InChI key

PYLWMHQQBFSUBP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Fluorobenzene is generally used as a solvent in synthetic chemistry.[1][2] It can also be used as an aryl fluoride precursor in the nickel-catalyzed cross-coupling reactions.[3][4]

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Dam. 1 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

5.0 °F - closed cup

Flash Point(C)

-15 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nickel-catalyzed Suzuki-Miyaura reaction of aryl fluorides.
Tobisu, Mamoru et al.
Journal of the American Chemical Society, 133(48), 19505-19511 (2011)
Reactions of a cyclic rhodium carbenoid with aromatic compounds and vinyl ethers.
Pirrung, Michael C et al.
The Journal of Organic Chemistry, 60(7), 2112-2124 (1995)
Coordinatively unsaturated ruthenium phosphine half-sandwich complexes: Correlations to structure and reactivity.
Aneetha, Halikhedkar et al.
Organometallics, 21(24), 5334-5346 (2002)
Catalytic C- C bond formation through selective activation of C- F bonds.
Bohm, Volker PW et al.
Angewandte Chemie (International Edition in English), 40(18), 3387-3389 (2001)
Fidel Ortega-Gavilán et al.
Food chemistry, 322, 126743-126743 (2020-04-14)
Sensory properties are critical characteristics that determine quality and can be evaluated by trained tasting panels. The panels function as multi-sensor measuring instrument and need the use of reference materials (RMs) for training. The homogeneity between units packaged from a

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service