![Chloro(dimethyl)octylsilane 97%](/deepweb/assets/sigmaaldrich/product/structures/396/605/8b3dddb4-bae6-4660-8b72-dd8eb2b78cb4/640/8b3dddb4-bae6-4660-8b72-dd8eb2b78cb4.png)
Chloro(dimethyl)octylsilane
![Chloro(dimethyl)octylsilane 97%](/deepweb/assets/sigmaaldrich/product/structures/396/605/8b3dddb4-bae6-4660-8b72-dd8eb2b78cb4/640/8b3dddb4-bae6-4660-8b72-dd8eb2b78cb4.png)
Synonym(s):
Dimethyloctylchlorosilane, Octyldimethylchlorosilane
Linear Formula:
CH3(CH2)7Si(CH3)2Cl
Products
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing