Skip to Content
Merck
All Photos(2)

Key Documents

242470

Sigma-Aldrich

7-Bromoheptanenitrile

90%

Synonym(s):

6-Cyanohexyl bromide

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
€101.00

€101.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
€101.00

About This Item

Linear Formula:
Br(CH2)6CN
CAS Number:
Molecular Weight:
190.08
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€101.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

90%

form

liquid

refractive index

n20/D 1.475 (lit.)

bp

140-141 °C/14 mmHg (lit.)

density

1.265 g/mL at 25 °C (lit.)

functional group

bromo
nitrile

SMILES string

BrCCCCCCC#N

InChI

1S/C7H12BrN/c8-6-4-2-1-3-5-7-9/h1-6H2

InChI key

HVVQSKCGHAPHMV-UHFFFAOYSA-N

Application

7-Bromoheptanenitrile was used in the synthesis of (±)-15-deoxyspergualin, a new immunosuppressive agent[1].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A new efficient synthesis of the immunosuppressive agent (?)-15-deoxyspergualin.
Tetrahedron, 57(14), 2757-2760 (2001)
Franziska Kupke et al.
Scientific reports, 6, 37631-37631 (2016-11-25)
Isothiocyanates are the most intensively studied breakdown products of glucosinolates from Brassica plants and well recognized for their pleiotropic effects against cancer but also for their genotoxic potential. However, knowledge about the bioactivity of glucosinolate-borne nitriles in foods is very

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service