Skip to Content
Merck
All Photos(1)

Key Documents

267880

Sigma-Aldrich

1,2-Bis(chlorodimethylsilyl)ethane

96%

Synonym(s):

1,1,4,4-Tetramethyl-1,4-dichloro-disilethylene, 2,5-Dichloro-2,5-dimethyl-2,5-disilahexane

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€84.30

€84.30


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
€84.30

About This Item

Linear Formula:
[-CH2Si(CH3)2Cl]2
CAS Number:
Molecular Weight:
215.27
Beilstein:
1920141
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

€84.30


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

96%

form

solid

bp

198 °C/734 mmHg (lit.)

mp

35-38 °C (lit.)

SMILES string

C[Si](C)(Cl)CC[Si](C)(C)Cl

InChI

1S/C6H16Cl2Si2/c1-9(2,7)5-6-10(3,4)8/h5-6H2,1-4H3

InChI key

VGQOKOYKFDUPPJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

149.0 °F - closed cup

Flash Point(C)

65 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrahedron Letters, 31, 4289-4289 (1990)
Tetrahedron Letters, 22, 1787-1787 (1981)
Jeyeong Lee et al.
Dalton transactions (Cambridge, England : 2003), 49(42), 15002-15008 (2020-10-24)
The self-assembly of Pd(NO3)2 with L (L = 1,2-bis(dimethyl(pyridin-3-yl)silyl)ethane) gives rise to [PdL2](NO3)2 in high yields. Anion exchange of [PdL2](NO3)2 with X- (X- = BF4-, ClO4-, and PF6-) changes the skeleton into a cage of [(X)@Pd2L4](X)3. Successive anion exchange of
Nikos Petzetakis et al.
Nature communications, 6, 7529-7529 (2015-06-25)
Free-volume of polymers governs transport of penetrants through polymeric films. Control over free-volume is thus important for the development of better membranes for a wide variety of applications such as gas separations, pharmaceutical purifications and energy storage. To date, methodologies

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service