Skip to Content
Merck
All Photos(4)

Key Documents

703567

Sigma-Aldrich

Isopropylmagnesium bromide solution

greener alternative

2.9 M in 2-methyltetrahydrofuran

Synonym(s):

1-Methylethylmagnesium bromide, 2-Propylmagnesium bromide, iso-Propylmagnesium bromide, Bromoisopropylmagnesium

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€162.00

€162.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
€162.00

About This Item

Linear Formula:
(CH3)2MgBr
CAS Number:
Molecular Weight:
147.30
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

€162.00


Check Cart for Availability

Request a Bulk Order

form

liquid

Quality Level

reaction suitability

reaction type: Grignard Reaction

greener alternative product characteristics

Safer Solvents and Auxiliaries
Use of Renewable Feedstocks
Inherently Safer Chemistry for Accident Prevention
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

concentration

2.9 M in 2-methyltetrahydrofuran
40 % (w/w)

density

1.112 g/mL at 25 °C

greener alternative category

SMILES string

CC(C)[Mg]Br

InChI

1S/C3H7.BrH.Mg/c1-3-2;;/h3H,1-2H3;1H;/q;;+1/p-1

InChI key

UCJIVFREPMUZDC-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product, 2.9 M in 2-methyltetrahydrofuran aligns with Safer Solvents and Auxiliaries, Use of Renewable Feedstocks and Inherently Safer Chemistry for Accident Prevention. Click here for more information.

Application

Grignard reagent in greener solvent, 2-methyltetrahydrofuran (2-MeTHF)

2-Methyltetrahydrofuran (2-MeTHF): A Biomass-Derived Solvent with Broad Application in Organic Chemistry

Legal Information

Product of Albemarle US Inc

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - Water-react 2

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 2

Flash Point(F)

-13.0 °F

Flash Point(C)

-25 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service