Skip to Content
Merck
All Photos(1)

Key Documents

187917

Sigma-Aldrich

Cyclobutanemethanol

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C4H7CH2OH
CAS Number:
Molecular Weight:
86.13
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form:
liquid
Assay:
99%

Assay

99%

form

liquid

refractive index

n20/D 1.446 (lit.)

bp

143-144 °C (lit.)

density

0.913 g/mL at 25 °C (lit.)

SMILES string

OCC1CCC1

InChI

1S/C5H10O/c6-4-5-2-1-3-5/h5-6H,1-4H2

InChI key

WPOPOPFNZYPKAV-UHFFFAOYSA-N

General description

Oxidation of cyclobutanemethanol in the presence of lead tetraacetate and lead tetraacetate/metal chloride has been investigated[1].

Application

Cyclobutanemethanol was used in the synthesis of bicyclic lactone[2].

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

104.0 °F - closed cup

Flash Point(C)

40 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The lead tetraacetate reaction of alcohols containing a small ring. Part II. Cyclobutane-methanols and cyclopropane-ethanols.
Mihailovic ML, et al.
Helvetica Chimica Acta, 59(2), 475-486 (1976)
Total synthesis of (-)-pestalotiopsin A.
Ken-ichi Takao et al.
Angewandte Chemie (International ed. in English), 47(18), 3426-3429 (2008-03-28)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service