Skip to Content
Merck
All Photos(1)

Key Documents

129402

Sigma-Aldrich

3-Methyl-3-buten-1-ol

97%

Synonym(s):

3-Isopentenyl alcohol, Isoprenol

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€64.20
500 ML
€135.00

€64.20


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
€64.20
500 ML
€135.00

About This Item

Linear Formula:
CH2=C(CH3)CH2CH2OH
CAS Number:
Molecular Weight:
86.13
Beilstein:
1071239
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€64.20


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.433 (lit.)

bp

130-132 °C (lit.)

density

0.853 g/mL at 25 °C (lit.)

functional group

hydroxyl

SMILES string

CC(=C)CCO

InChI

1S/C5H10O/c1-5(2)3-4-6/h6H,1,3-4H2,2H3

InChI key

CPJRRXSHAYUTGL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3-Methyl-3-buten-1-ol was used to study the rate coefficient for the gas-phase reaction of OH radical with 3-methyl-3-buten-1-ol[1].

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

107.6 °F - closed cup

Flash Point(C)

42 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pablo M Cometto et al.
The journal of physical chemistry. A, 112(19), 4444-4450 (2008-04-25)
Rate coefficients for the gas-phase reactions of OH radicals with four unsaturated alcohols, 3-methyl-3-buten-1-ol (k1), 2-buten-1-ol (k2), 2-methyl-2-propen-1-ol (k3) and 3-buten-1-ol (k4), were measured using two different techniques, a conventional relative rate method and the pulsed laser photolysis-laser induced fluorescence
Tian-He Wang et al.
Huan jing ke xue= Huanjing kexue, 32(12), 3599-3605 (2012-04-04)
Multiphase acid-catalyzed oxidation with hydrogen peroxide (H2O2) has been suggested recently to be a potential route to SOA formation, but the kinetics and chemical mechanism of this process have not been well-known yet. In this work, the uptake of 3-methyl-3-buten-1-ol
[Penetration of primary and tertiary alcohol isomers through rubber and isolated skin].
A L Mikhaleva et al.
Gigiena truda i professional'nye zabolevaniia, (11)(11), 46-47 (1982-01-01)

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service