Skip to Content
Merck
All Photos(1)

Key Documents

159263

Sigma-Aldrich

(1-Ethoxyethylidene)malononitrile

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C(OC2H5)=C(CN)2
CAS Number:
Molecular Weight:
136.15
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Assay:
98%

Assay

98%

mp

90-92 °C (lit.)

functional group

ether
nitrile

SMILES string

CCO\C(C)=C(\C#N)C#N

InChI

1S/C7H8N2O/c1-3-10-6(2)7(4-8)5-9/h3H2,1-2H3

InChI key

BOSVWXDDFBSSIZ-UHFFFAOYSA-N

General description

(1-Ethoxyethylidene)malononitrile is also referred as 2-(1-ethoxyethylidene)malononitrile.[1]

Application

(1-Ethoxyethylidene)malononitrile was used in the synthesis of benzyl 5-amino-4-cyano-3-methyl-1H pyrazole-1-carboxylate[1] and 2-amino-6-mercaptopyridine-3,5-dicarbonitrile derivatives.[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yaojun Gao et al.
Journal of combinatorial chemistry, 12(1), 69-74 (2009-12-05)
A solid-phase synthesis of 5-aminopyrazole has been developed and applied to the preparation of pyrazolo[5,1-d][1,2,3,5]tetrazine-4(3H)-ones. In this strategy, a one-pot reaction from 5-aminopyrazoles to the pyrazolo[5,1-d][1,2,3,5]tetrazine-4(3H)-ones which provided the compounds in good yields was demonstrated. Using this synthetic strategy, we
Diaa A Ibrahim et al.
European journal of medicinal chemistry, 46(12), 5825-5832 (2011-10-18)
The design and synthesis of a small library of 4-aminopyrido[2,3-d]pyrimidine derivatives is reported. The potential activity of these compounds as CDK2/Cyclin A, CDK4/Cyclin D, EGFR and anti-tumor was evaluated by cytotoxicity studies in A431a, SNU638b, HCT116 and inhibition of CDK2-Cyclin

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service