Skip to Content
Merck
All Photos(1)

Key Documents

M54201

Sigma-Aldrich

2-(Methylthio)aniline

97%

Synonym(s):

2-(Methylmercapto)aniline, 2-Aminothioanisole

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€156.00

€156.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
€156.00

About This Item

Linear Formula:
CH3SC6H4NH2
CAS Number:
Molecular Weight:
139.22
Beilstein:
386211
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€156.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

refractive index

n20/D 1.624 (lit.)

bp

234 °C (lit.)

density

1.111 g/mL at 25 °C (lit.)

SMILES string

CSc1ccccc1N

InChI

1S/C7H9NS/c1-9-7-5-3-2-4-6(7)8/h2-5H,8H2,1H3

InChI key

WBRPQQSADOCKCH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

>235.4 °F - closed cup

Flash Point(C)

> 113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Manisha Das et al.
Dalton transactions (Cambridge, England : 2003), 48(4), 1292-1313 (2019-01-05)
Rational ligand design approaches allowed {Cu2(μ-OH/OMe)} cores to be accommodated within μ-phenoxido bis(tetradentate) and μ-phenoxido bis(tridentate) ligands having thioether donors. The complexes [Cu2(μ-H2L1)(μ-OH)](ClO4)2·2H2O (1), [Cu2(μ-L2)(μ-OH)(OH2)](ClO4)2 (2a) and [Cu2(μ-L2)(μ-OCH3)(OH2)](ClO4)2 (2b) were obtained from an N2O3S2 donor set bearing the H3L1 ligand
Takashi Ikawa et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 26(19), 4320-4332 (2020-01-03)
Benzynes were selectively generated in situ from phenols and trapped regioselectively with potassium hexamethyldisilazide to form primary anilines following acidic workup. The direct conversion of a phenolic hydroxyl group into a free amino group is a useful method for the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service