Skip to Content
Merck
All Photos(2)

Key Documents

E6200

Sigma-Aldrich

(Ethoxymethylene)malononitrile

98%

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€32.90
100 G
€71.40

€32.90


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
€32.90
100 G
€71.40

About This Item

Linear Formula:
C2H5OCH=C(CN)2
CAS Number:
Molecular Weight:
122.12
Beilstein:
1634241
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€32.90


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

bp

160 °C/12 mmHg (lit.)

mp

64-66 °C (lit.)

SMILES string

CCO\C=C(\C#N)C#N

InChI

1S/C6H6N2O/c1-2-9-5-6(3-7)4-8/h5H,2H2,1H3

InChI key

OEICGMPRFOJHKO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Caution

This product is known to be a strong sensitizing agent

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eszter Szennyes et al.
Molecules (Basel, Switzerland), 25(3) (2020-02-12)
Despite the substantial interest in C-glycosyl heterocycles as mimetics of biologically active native glycans, the appearance of C-glycopyranosyl derivatives of six-membered heterocycles, both in synthetic and biological contexts, is rather scarce. As part of our ongoing research program aimed at

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service