Skip to Content
Merck
All Photos(1)

Key Documents

152668

Sigma-Aldrich

3-Amino-2,5-dichlorobenzoic acid

95%

Synonym(s):

Chloramben

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€284.00

€284.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€284.00

About This Item

Linear Formula:
H2NC6H2(Cl)2CO2H
CAS Number:
Molecular Weight:
206.03
Beilstein:
2365906
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22

€284.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

95%

reaction suitability

reaction type: solution phase peptide synthesis

mp

194-197 °C (dec.) (lit.)

application(s)

peptide synthesis

SMILES string

Nc1cc(Cl)cc(C(O)=O)c1Cl

InChI

1S/C7H5Cl2NO2/c8-3-1-4(7(11)12)6(9)5(10)2-3/h1-2H,10H2,(H,11,12)

InChI key

HSSBORCLYSCBJR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S C Fang et al.
Journal of pharmacokinetics and biopharmaceutics, 8(6), 583-597 (1980-12-01)
The acidic compounds, such as phenoxyacetic acids, substituted benzoic acids, or acetylsalicylic acid, were found to bind to bovine serum albumin (BSA). Among phenoxyacetic acids, the binding affinity to BSA was highest for 2,4,5-trichlorophenoxyacetic acid (2,4,5-T), which was approximately 4-
Field persistence studies with triallate and trifluralin both singly and in combination with chloramben.
A E Smith et al.
Bulletin of environmental contamination and toxicology, 29(2), 240-242 (1982-08-01)
[Establishment of the maximum permissible concentration of amiben in the water of reservoirs].
S Iu Buslovich et al.
Gigiena i sanitariia, (4)(4), 74-75 (1980-04-01)
[Methods of controlling the harmful substance content in the air of a work area in amiben manufacture].
I I Pilenkova et al.
Gigiena truda i professional'nye zabolevaniia, (10)(10), 50-51 (1980-10-01)
Ilaria Massarelli et al.
European journal of medicinal chemistry, 44(9), 3658-3664 (2009-03-11)
A dataset comprising 55 chemicals with hepatocarcinogenic potency indices was collected from the Carcinogenic Potency Database with the aim of developing QSAR models enabling prediction of the above unwanted property for New Chemical Entities. The dataset was rationally split into

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service