Skip to Content
Merck
All Photos(2)

Key Documents

33941

Supelco

Thionazin

PESTANAL®, analytical standard

Synonym(s):

O,O-Diethyl O-(2-pyrazinyl) phosphorothioate

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
€288.00

€288.00


Estimated to ship on17 May 2025


Request a Bulk Order

Select a Size

Change View
50 MG
€288.00

About This Item

Empirical Formula (Hill Notation):
C8H13N2O3PS
CAS Number:
Molecular Weight:
248.24
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

€288.00


Estimated to ship on17 May 2025


Request a Bulk Order

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

CCOP(=S)(OCC)Oc1cnccn1

InChI

1S/C8H13N2O3PS/c1-3-11-14(15,12-4-2)13-8-7-9-5-6-10-8/h5-7H,3-4H2,1-2H3

InChI key

IRVDMKJLOCGUBJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Thionazin is an amber coloured liquid, which finds applications as a soil fungicide, insecticide and nematicide[1][2]

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sittig's Handbook of Pesticides and Agricultural Chemicals (2013)
Science of Synthesis: Houben-Weyl Methods of Molecular Transformations, 16 (2014)
G Fiori et al.
Schweizerische medizinische Wochenschrift, 117(11), 399-401 (1987-03-14)
Among the insecticides, organophosphorous compounds are the agents most involved in deliberate and involuntary intoxications. The toxic effect is mainly due to the inhibition of cholinesterase at the cholinergic synapses. Two cases of severe involuntary intoxication are described which were
F Sepulcri et al.
Schweizerische medizinische Wochenschrift, 117(13), 487-490 (1987-03-28)
We report on a case of severe intoxication in an 18-year-old female who had ingested an insecticide with suicidal intent. The insecticide (thionacine) is a cholinesterase inhibitor used in gardening and agriculture. The resources of modern intensive care permitted successful
Maya Kochman et al.
Journal of chromatography. A, 1129(1), 95-104 (2006-07-13)
A new approach of flow modulation comprehensive two-dimensional gas chromatography-mass spectrometry (GC x GC-MS) with supersonic molecular beam (SMB) and a quadrupole mass analyzer is presented. Flow modulation uniquely enables GC x GC-MS to be achieved even with the limited

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service