Skip to Content
Merck
All Photos(1)

Key Documents

339598

Sigma-Aldrich

4-Amino-3-hydroxybenzoic acid

97%

Synonym(s):

3-Hydroxy-4-aminobenzoic acid, 3-Hydroxy-PABA

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€67.30
25 G
€122.00

€67.30


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€67.30
25 G
€122.00

About This Item

Linear Formula:
H2NC6H3(OH)CO2H
CAS Number:
Molecular Weight:
153.14
Beilstein:
509859
MDL number:
UNSPSC Code:
12352106
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.22

€67.30


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

solid

reaction suitability

reaction type: solution phase peptide synthesis

mp

211-215 °C (lit.)

application(s)

peptide synthesis

SMILES string

Nc1ccc(cc1O)C(O)=O

InChI

1S/C7H7NO3/c8-5-2-1-4(7(10)11)3-6(5)9/h1-3,9H,8H2,(H,10,11)

InChI key

NFPYJDZQOKCYIE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Kerim Babaoglu et al.
Nature chemical biology, 2(12), 720-723 (2006-10-31)
Fragment-based screens test multiple low-molecular weight molecules for binding to a target. Fragments often bind with low affinities but typically have better ligand efficiencies (DeltaG(bind)/heavy atom count) than traditional screening hits. This efficiency, combined with accompanying atomic-resolution structures, has made
Jinkyu Park et al.
Journal of hazardous materials, 371, 243-252 (2019-03-11)
Carboxylate-functionalized organic nanocrystals (ONCs) derived from perylene diimide or naphthalene diimide were synthesized and carefully characterized as novel high-capacity uranium (U(VI)) sorbents. Adsorption studies using uranyl ions demonstrated that the carboxyl and hydroxyl groups on the surface of the ONCs
Falicia Goh et al.
Microbial cell factories, 19(1), 71-71 (2020-03-21)
Notonesomycin A is a 32-membered bioactive glycosylated macrolactone known to be produced by Streptomyces aminophilus subsp. notonesogenes 647-AV1 and S. aminophilus DSM 40186. In a high throughput antifungal screening campaign, we identified an alternative notonesomycin A producing strain, Streptomyces sp.
Chao-Bin Xue et al.
Bioorganic & medicinal chemistry, 15(5), 2006-2015 (2007-01-30)
Phenoloxidase (PO), also known as tyrosinase, is a key enzyme in insect development, responsible for catalyzing the hydroxylation of tyrosine into o-diphenols and the oxidation of o-diphenols into o-quinones. Inhibition of PO may provide a basis for novel environmentally friendly

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service