Skip to Content
Merck
All Photos(4)

Key Documents

133698

Sigma-Aldrich

4-Chlorobenzenesulfonyl chloride

97%

Synonym(s):

p-Chlorobenzenesulfonyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H4SO2Cl
CAS Number:
Molecular Weight:
211.07
Beilstein:
511583
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39092438
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

97%

form

solid

bp

141 °C/15 mmHg (lit.)

mp

50-52 °C (lit.)

functional group

chloro

SMILES string

Clc1ccc(cc1)S(Cl)(=O)=O

InChI

1S/C6H4Cl2O2S/c7-5-1-3-6(4-2-5)11(8,9)10/h1-4H

InChI key

ZLYBFBAHAQEEQQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4-Chlorobenzenesulfonyl chloride was used in the synthesis of precursors for the 3,4-pyridyne generation[1].

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

226.4 °F - closed cup

Flash Point(C)

108 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation of functionalized 3, 4-pyridynes via 2-magnesiated diaryl sulfonates.
Lin W, et al.
Tetrahedron, 63(13), 2787-2797 (2007)
Tsurng-Juhn Huang et al.
European journal of medicinal chemistry, 90, 428-435 (2014-12-03)
Hepatitis B virus (HBV) is a causative reagent that frequently causes progressive liver diseases, leading to the development of acute, chronic hepatitis, cirrhosis, and eventually hepatocellular carcinoma (HCC). Despite several antiviral drugs including interferon-α and nucleotide derivatives are approved for

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service