Skip to Content
Merck
All Photos(1)

Key Documents

150525

Sigma-Aldrich

trans-β-Styrenesulfonyl chloride

97%

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
€357.00

€357.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
€357.00

About This Item

Linear Formula:
C6H5CH=CHSO2Cl
CAS Number:
Molecular Weight:
202.66
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€357.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

mp

85-88 °C (lit.)

functional group

phenyl

SMILES string

[H]\C(=C(\[H])S(Cl)(=O)=O)c1ccccc1

InChI

1S/C8H7ClO2S/c9-12(10,11)7-6-8-4-2-1-3-5-8/h1-7H/b7-6+

InChI key

ONWRSBMOCIQLRK-VOTSOKGWSA-N

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zoon H Ryu et al.
International journal of molecular sciences, 9(12), 2639-2657 (2009-03-31)
Additional specific rates of solvolysis have been determined, mainly in fluoroalcohol containing solvents, for benzenesulfonyl chloride (1) and p-nitrobenzene-sulfonyl chloride (2). For trans-beta-styrenesulfonyl chloride (3), a study has been carried out in 43 pure and binary solvents, covering a wide

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service