Skip to Content
Merck
All Photos(1)

Key Documents

272248

Sigma-Aldrich

4-Nitrobenzenesulfonyl chloride

97%

Synonym(s):

4-Nitrobenzene-1-sulfonyl chloride, 4-Nitrobenzenesulfonic acid chloride, 4-Nitrobenzenesulfonyl chloride, 4-Nitrophenyl-1-sulfonyl chloride, 4-Nitrophenylsulfonyl chloride, Nosyl chloride, p-Nitrobenzenesulfonyl chloride, p-Nitrophenylsulfonyl chloride, p-Nosyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H4SO2Cl
CAS Number:
Molecular Weight:
221.62
Beilstein:
746543
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

97%

mp

66-70 °C (lit.)

functional group

nitro

SMILES string

[O-][N+](=O)c1ccc(cc1)S(Cl)(=O)=O

InChI

1S/C6H4ClNO4S/c7-13(11,12)6-3-1-5(2-4-6)8(9)10/h1-4H

InChI key

JXRGUPLJCCDGKG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4-Nitrobenzenesulfonyl chloride was used in the synthesis of
  • [Cu(bipy)2Cl](nbs) (1) (bipy = 2,2′-bipyridine, nbs = 4-nitrobenzenesulfonate)[1]
  • hydroxylamines of sulfadiazine and sulfamethoxazole[2]
  • solution phase peptide using N-nosyl-α-amino acids[3].

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Repr. 2 - Skin Sens. 1A

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrahedron, 63, 8164-8164 (2007)
M J Rieder et al.
The Journal of pharmacology and experimental therapeutics, 244(2), 724-728 (1988-02-01)
Among the most serious side effects of sulfonamides are hypersensitivity reactions, the pathogenesis of which has been suggested to be mediated by reactive metabolites. We have previously demonstrated dose-related covalent binding and toxicity of reactive intermediates of sulfonamides generated by
Jason Jones et al.
Analytical chemistry, 92(4), 3483-3491 (2020-01-29)
For decades, there has been immense progress in miniaturizing analytical methods based on electrophoresis to improve sensitivity and to reduce sample volumes, separation times, and/or equipment cost and space requirements, in applications ranging from analysis of biological samples to environmental
Mahboubeh A Sharif et al.
Acta chimica Slovenica, 59(2), 289-293 (2012-06-01)
[Cu(bipy)2Cl](nbs) (1) (bipy = 2,2'-bipyridine, nbs = 4-nitrobenzenesulfonate) was obtained from the reaction of 4-nitrobenzenesulfonyl chloride and 2-amine-4-methylopyridine with CuCl2 in the presence of 2,2'-bipyridine and characterized by elemental analysis, IR spectra and X-ray single-crystal diffraction. The asymmetric unit of
Balázs Pásztói et al.
Polymers, 12(11) (2020-11-01)
Endfunctional polymers possess significant industrial and scientific importance. Sulfonyl endgroups, such as tosyl and nosyl endfunctionalities, due their ease of substitution are highly desired for a variety of polymer structures. The sulfonylation of hydroxyl-terminated polyisobutylene (PIB-OH), a chemically and thermally

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service