Skip to Content
Merck
All Photos(1)

Documents

111856

Sigma-Aldrich

Propylcyclohexane

99%

Synonym(s):

n-Propylcyclohexane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H11CH2CH2CH3
CAS Number:
Molecular Weight:
126.24
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

8.7 mmHg ( 37.7 °C)

Quality Level

Assay

99%

autoignition temp.

478 °F

refractive index

n20/D 1.436 (lit.)

bp

155 °C (lit.)

density

0.793 g/mL at 25 °C (lit.)

SMILES string

CCCC1CCCCC1

InChI

1S/C9H18/c1-2-6-9-7-4-3-5-8-9/h9H,2-8H2,1H3

InChI key

DEDZSLCZHWTGOR-UHFFFAOYSA-N

Application

Propylcyclohexane is a component of the surrogate biokerosene model fuel. It has been used in the kinetic studies of kerosene Jet A-1 oxidation. It has also been used to assay kerosene/rapeseed oil methyl ester (RME) mixture (80/20, mol/mol; biokerosene).

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

95.0 °F - closed cup

Flash Point(C)

35 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Philippe Dagaut et al.
The journal of physical chemistry. A, 111(19), 3992-4000 (2007-01-27)
The kinetics of oxidation of kerosene Jet A-1 and a kerosene/rapeseed oil methyl ester (RME) mixture (80/20, mol/mol) (biokerosene) was studied experimentally in a jet-stirred reactor at 10 atm and constant residence time, over the temperature range 740-1200 K, and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service