Skip to Content
Merck
All Photos(1)

Key Documents

110493

Sigma-Aldrich

1,1-Dimethylcyclohexane

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H10(CH3)2
CAS Number:
Molecular Weight:
112.21
Beilstein:
1900304
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Assay:
99%

vapor pressure

42 mmHg ( 37.7 °C)

Quality Level

Assay

99%

refractive index

n20/D 1.428 (lit.)

bp

118-120 °C (lit.)

density

0.777 g/mL at 25 °C (lit.)

SMILES string

CC1(C)CCCCC1

InChI

1S/C8H16/c1-8(2)6-4-3-5-7-8/h3-7H2,1-2H3

InChI key

QEGNUYASOUJEHD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,1-Dimethylcyclohexane is an additive that facilitates mild-pressure handling of natural-gas hydrate.

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Sepideh Khalatbari-Limaki et al.
Iranian journal of microbiology, 12(6), 616-624 (2021-02-23)
Foodborne pathogens are among the serious problems all around the world and thus a novel and natural strategy to control and to inhibit such pathogens is highly demanded nowadays. The aim of this study was to isolate a specific bacteriophage
Babak Pakbin et al.
Antibiotics (Basel, Switzerland), 10(11) (2021-11-28)
The emergence of multi-drug resistant E. coli is an important matter of increasing considerable concern to global public health. The aim of this study was to investigate the incidence, antibiotic resistance pattern and phylogroups of E. coli isolates obtained from
Large pressure depression of methane hydrate by adding 1, 1-dimethylcyclohexane.
Chemical Engineering Science, 60(11), 3117-3119 (2005)
Enriqueta Garcia-Gutierrez et al.
Scientific reports, 10(1), 3738-3738 (2020-03-01)
Nisin P is a natural nisin variant, the genetic determinants for which were previously identified in the genomes of two Streptococcus species, albeit with no confirmed evidence of production. Here we describe Streptococcus agalactiae DPC7040, a human faecal isolate, which
Lady D K T Prazeres et al.
Oxidative medicine and cellular longevity, 2019, 1983137-1983137 (2019-12-13)
Ethnomedicinal studies in the Amazon community and in the Northeast region of Brazil highlight the use of Libidibia ferrea fruits for the treatment of gastric problems. However, there are no data in the literature of this pharmacological activity. Thus, the

Articles

Dehydrated culture media for cultivating Shigella come in granules that dissolve in water. Granulation of the media ensures homogenous distribution, which avoids clumping even in warm and humid conditions. 

Shigella spp. are predominantly connected to humans and are rarely isolated from other animals, with the exception of primates.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service