Merck
  • Early insights into the interactions of different β-lactam antibiotics and β-lactamase inhibitors against soluble forms of Acinetobacter baumannii PBP1a and Acinetobacter sp. PBP3.
Early insights into the interactions of different β-lactam antibiotics and β-lactamase inhibitors against soluble forms of Acinetobacter baumannii PBP1a and Acinetobacter sp. PBP3.
Antimicrobial agents and chemotherapy (2012-08-22)
Krisztina M Papp-Wallace, Baui Senkfor, Julian Gatta, Weirui Chai, Magdalena A Taracila, Veerabahu Shanmugasundaram, Seungil Han, Richard P Zaniewski, Brian M Lacey, Andrew P Tomaras, Marion J Skalweit, Michael E Harris, Louis B Rice, John D Buynak, Robert A Bonomo
ZUSAMMENFASSUNG
Acinetobacter baumannii is an increasingly problematic pathogen in United States hospitals. Antibiotics that can treat A. baumannii are becoming more limited. Little is known about the contributions of penicillin binding proteins (PBPs), the target of β-lactam antibiotics, to β-lactam-sulbactam susceptibility and β-lactam resistance in A. baumannii. Decreased expression of PBPs as well as loss of binding of β-lactams to PBPs was previously shown to promote β-lactam resistance in A. baumannii. Using an in vitro assay with a reporter β-lactam, Bocillin, we determined that the 50% inhibitory concentrations (IC(50)s) for PBP1a from A. baumannii and PBP3 from Acinetobacter sp. ranged from 1 to 5 μM for a series of β-lactams. In contrast, PBP3 demonstrated a narrower range of IC(50)s against β-lactamase inhibitors than PBP1a (ranges, 4 to 5 versus 8 to 144 μM, respectively). A molecular model with ampicillin and sulbactam positioned in the active site of PBP3 reveals that both compounds interact similarly with residues Thr526, Thr528, and Ser390. Accepting that many interactions with cell wall targets are possible with the ampicillin-sulbactam combination, the low IC(50)s of ampicillin and sulbactam for PBP3 may contribute to understanding why this combination is effective against A. baumannii. Unraveling the contribution of PBPs to β-lactam susceptibility and resistance brings us one step closer to identifying which PBPs are the best targets for novel β-lactams.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Sulbactam
Sulbactam, European Pharmacopoeia (EP) Reference Standard