Merck
Expedient synthesis of sulfinamides from sulfonyl chlorides.
The Journal of organic chemistry (2007-01-16)
Michael Harmata, Pinguan Zheng, Chaofeng Huang, Maria G Gomes, Weijiang Ying, Kanok-On Rayanil, Kanok-On Ranyanil, Gayatri Balan, Nathan L Calkins
ZUSAMMENFASSUNG
Sulfinamides were synthesized from sulfonyl chlorides using a procedure involving in situ reduction of sulfonyl chlorides. The reaction is broad in scope and easy to perform.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Sulfurylchlorid, 97%
Sigma-Aldrich
Sulfurylchlorid -Lösung, 1.0 M in methylene chloride