Merck
Non-oncogene Addiction to SIRT3 Plays a Critical Role in Lymphomagenesis.
Cancer cell (2019-06-12)
Meng Li, Ying-Ling Chiang, Costas A Lyssiotis, Matthew R Teater, Jun Young Hong, Hao Shen, Ling Wang, Jing Hu, Hui Jing, Zhengming Chen, Neeraj Jain, Cihangir Duy, Sucharita J Mistry, Leandro Cerchietti, Justin R Cross, Lewis C Cantley, Michael R Green, Hening Lin, Ari M Melnick
ZUSAMMENFASSUNG
Diffuse large B cell lymphomas (DLBCLs) are genetically heterogeneous and highly proliferative neoplasms derived from germinal center (GC) B cells. Here, we show that DLBCLs are dependent on mitochondrial lysine deacetylase SIRT3 for proliferation, survival, self-renewal, and tumor growth in vivo regardless of disease subtype and genetics. SIRT3 knockout attenuated B cell lymphomagenesis in VavP-Bcl2 mice without affecting normal GC formation. Mechanistically, SIRT3 depletion impaired glutamine flux to the TCA cycle via glutamate dehydrogenase and reduction in acetyl-CoA pools, which in turn induce autophagy and cell death. We developed a mitochondrial-targeted class I sirtuin inhibitor, YC8-02, which phenocopied the effects of SIRT3 depletion and killed DLBCL cells. SIRT3 is thus a metabolic non-oncogene addiction and therapeutic target for DLBCLs.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-Histon H3.3-Antikörper, from rabbit, purified by affinity chromatography
Preise und Verfügbarkeit sind derzeit nicht verfügbar.