Merck
V900111
Sigma-Aldrich
Kaliumcyanat
Vetec, reagent grade, 96%
About This Item
Lineare Formel:
KOCN
CAS-Nummer:
Molekulargewicht:
81.12
Beilstein:
3594798
EG-Nummer:
MDL-Nummer:
UNSPSC-Code:
12352300
PubChem Substanz-ID:
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Qualität
reagent grade
Produktlinie
Vetec
Assay
96%
Form
powder, crystals or chunks
Dichte
2.056 g/mL at 25 °C (lit.)
SMILES String
[K+].[O-]C#N
InChI
1S/CHNO.K/c2-1-3;/h3H;/q;+1/p-1
InChIKey
GKKCIDNWFBPDBW-UHFFFAOYSA-M
Rechtliche Hinweise
Vetec is a trademark of Merck KGaA, Darmstadt, Germany
Piktogramme
Exclamation mark
Signalwort
Warning
Gefahreneinstufungen
Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2
Lagerklassenschlüssel
11 - Combustible Solids
WGK
WGK 1
Flammpunkt (°F)
Not applicable
Flammpunkt (°C)
Not applicable
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Die Dokumentenbibliothek aufrufen
Jeroen N Stoop et al.
PloS one, 9(7), e102163-e102163 (2014-07-16)
The immune response to post-translationally modified antigens is a key characteristic of rheumatoid arthritis. Carbamylation is such a posttranslational modification. Recently, we demonstrated that autoantibodies recognizing carbamylated proteins are present in sera of rheumatoid arthritis. The molecular mechanisms underlying the
Jirí Verner et al.
Molecules (Basel, Switzerland), 11(1), 34-42 (2007-10-27)
The reactivity of alicyclic ketazines in criss-cross cycloadditions was investigated. They react with potassium cyanate and ammonium thiocyanate in the presence of acetic acid to form spirocyclic perhydro[1,2,4]triazolo[1,2-alpha][1,2,4]triazole-1,5-diones and perhydro[1,2,4]triazolo[1,2-alpha][1,2,4]triazole-1,5-dithiones, respectively, in relatively high yields.
E S Roberts et al.
Inhalation toxicology, 18(3), 159-167 (2006-01-10)
Hydrogen sulfide (H(2)S) is a naturally occurring gas that is also associated with several industries. The potential for widespread human inhalation exposure to this toxic gas is recognized as a public health concern. The nasal epithelium is particularly susceptible to
Grégoire Danger et al.
Amino acids, 42(6), 2331-2341 (2011-07-20)
The reaction of cyanate with C-terminal carboxyl groups of peptides in aqueous solution was considered as a potential pathway for the abiotic formation of peptide bonds under the condition of the primitive Earth. The catalytic effect of dicarboxylic acids on
Chang Liu et al.
Analytica chimica acta, 641(1-2), 59-63 (2009-04-28)
Electrochemical measurement of respiratory chain activity is a rapid and reliable screening for the toxicity on microorganisms. Here, we investigated in-vitro effects of toxin on Escherichia coli (E. coli) that was taken as a model microorganism incubated with ferricyanide. The