Fortfahren mit
509477
SGE gasdichte Spritzen, Luer-Lock
250F-LL-GT, volume 0.25 mL (250 μL), needle size (not included)
Synonym(e):
SPRITZE SGE 250μL, LUER LOCK
Anmelden zur Ansicht der Organisations- und Vertragspreise.
Größe auswählen
Ansicht ändern
Größe/SKUVerfügbarkeitPreis
Über diesen Artikel
UNSPSC Code:
41122001
NACRES:
NB.03
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten
material
glass syringe
needle size
(not included)
feature
without needle, Fixed Luer Lock
packaging
pkg of 1 ea
manufacturer/tradename
SGE 006230
volume
0.25 mL (250 μL)
application(s)
general analytical
General description
SGE Gas Tight syringes are constructed of borosilicate glass, PTFE plunger tips, and KEL-F Luer cones depending upon the experimentation process. SGE syringes have been a consistent choice for LC and GC equipment. An interference fit between the plunger tip and the barrel enhances the gas sealing properties of the SGE syringe. Plungers are interchangeable. SGE gas tight syringes can be autoclaved, but the plunger should be removed. SGE gastight Luer lock syringe can be effectively used for the introduction of samples to the gas sampling and injection system (GASIS) inlet.
  • Maximum inertness for liquid or gas samples
  • Suitable for manual disperser/diluter applications
  • High accuracy of dispensed volume
  • Wide variety of terminations
  • Less time consuming
  • SGE gas tight syringes also improves work flow analysis
Ähnliche Artikel vergleichen
Vollständigen Vergleich anzeigen
Unterschiede anzeigen
1 of 1
Dieser Artikel
509493509469509515
description
250F-LL-GT, volume 0.25 mL (250 μL), needle size (not included)
description
2.5MDF-LL-GT, volume 2500 μL, needle size (not included)
description
100F-LL-GT, volume 100 μL, needle size (not included)
description
25MDR-LL-GT, volume 25 mL, needle size (not included)
material
glass syringe
material
glass syringe
material
glass syringe
material
glass syringe
packaging
pkg of 1 ea
packaging
pkg of 1 ea
packaging
pkg of 1 ea
packaging
pkg of 1 ea
volume
0.25 mL (250 μL)
volume
2500 μL
volume
100 μL
volume
25 mL
manufacturer/tradename
SGE 006230
manufacturer/tradename
SGE 008425
manufacturer/tradename
SGE 005230
manufacturer/tradename
SGE 009462
application(s)
general analytical
application(s)
general analytical
application(s)
general analytical
application(s)
general analytical
feature
Fixed Luer Lock, without needle
feature
needle type Luer lock
feature
Fixed Luer Lock
feature
needle type Luer lock
Still not finding the right product?
Explore all of our products under SGE gasdichte Spritzen, Luer-Lock
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)