Fortfahren mit
27439
Schraubkappe
black polypropylene (Top Hat), PTFE/silicone, for use with 4 mL vial with 13-425 thread, pkg of 100 ea
Anmelden zur Ansicht der Organisations- und Vertragspreise.
Größe auswählen
Ansicht ändern
Größe/SKUVerfügbarkeitPreis
Über diesen Artikel
UNSPSC Code:
41122001
NACRES:
NB.43
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten
material
PTFE/silicone , black polypropylene (Top Hat)
feature
pre-assembled
packaging
pkg of 100 ea
fitting
top hat cap
application(s)
general analytical
compatibility
for use with 4 mL vial with 13-425 thread
General description
Preassembled closure and septa designed to provide a positive physical lock to assure that components arrive intact.
Application
PTFE-silicone-PTFE screw cap may be used for capping samples contained in a micro-insert vial, to be loaded on to GC/MS for biochemical lipid determination, of fat mass in Caenorhabditis Elegans.
Screw cap may be used for capping water samples in amber glass vial comprising of a polytetrafluoroethylene silicon septum for the analysis of chlorinated volatile organic compounds in water samples using headspace-solid-phase microextraction with gas chromatography (HS-SPME-GC).
Ähnliche Artikel vergleichen
Vollständigen Vergleich anzeigen
Unterschiede anzeigen
1 of 1
Dieser Artikel
27262BR130324BR130322
description
black polypropylene (Top Hat), PTFE/silicone, for use with 4 mL vial with 13-425 thread, pkg of 100 ea
description
black polypropylene hole cap, thread for 8-425, PTFE/silicone, septum thickness 1.5 mm, for use with 2 mL vial (standard opening), pkg of 100 ea
description
thread for GL W 54
description
thread for GL W 45
fitting
top hat cap
fitting
thread for 8-425
fitting
thread for GL W 54
fitting
thread for GL W 45
feature
pre-assembled
feature
closure type black polypropylene hole cap
feature
-
feature
-
material
black polypropylene (Top Hat), PTFE/silicone
material
PTFE/silicone , black polypropylene hole cap
material
polypropylene
material
polypropylene
compatibility
for use with 4 mL vial with 13-425 thread
compatibility
for use with 2 mL vial (standard opening)
compatibility
-
compatibility
-
packaging
pkg of 100 ea
packaging
pkg of 100 ea
packaging
pack of 1 ea
packaging
pack of 1 ea
application(s)
general analytical
application(s)
general analytical
application(s)
-
application(s)
-
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)