Fortfahren mit
22509
Supelco Ferrule Nut Adapter (fits Agilent injectors and non-MS detectors), knurled fingertight version
pkg of 2 ea
Anmelden zur Ansicht der Organisations- und Vertragspreise.
Über diesen Artikel
UNSPSC Code:
41121815
packaging
pkg of 2 ea
technique(s)
gas chromatography (GC): suitable using :Suitable
General description
These stainless steel adapters enable you to use general purpose and other 1/16 in. compression ferrules in Agilent GCs. Knurled head allows tightening with fingers, eliminating the need to locate the proper wrench.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)