Fortfahren mit
22003
Swagelok® Mutter, Ferrulen-Set vorn und hinten
SWAGELOK® 400-S, brass, 1/4 in. SWAGELOK
Anmelden zur Ansicht der Organisations- und Vertragspreise.
Größe auswählen
Ansicht ändern
Größe/SKUVerfügbarkeitPreis
Über diesen Artikel
UNSPSC Code:
41115717
NACRES:
SB.54
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten
material
brass
packaging
pkg of 10 ea
manufacturer/tradename
SWAGELOK® 400-S
technique(s)
gas chromatography (GC): suitable using :Suitable
fitting
1/4 in. SWAGELOK
General description
Swage tubing coupler has four parts namely the body, into which the tube is placed, two ferrules that is front ferrule and back ferrule and lastly a packing nut. While assembling a compression tube fitting, the metal tubing should be inspected properly to ensure that the surface is smooth. A nut is slided on to the tube fitting with the open side facing the end of the tubing. Followed by sliding the back ferrule with the wider part facing the nut. Further, sliding on the front ferrule with the small end of the cone facing the end of the tubing. Then inserting the tubing into the fitting and tighten up the nut/ferrule assembly onto the fitting.
Application
Swagelok® nut, front and back ferrule set maybe used for sampler construction from a single capillary gas chromatographic column, for collection of semi-volatile organic compounds through a diffusion denuder.
Legal Information
SWAGELOK is a registered trademark of Swagelok Company
Ähnliche Artikel vergleichen
Vollständigen Vergleich anzeigen
Unterschiede anzeigen
1 of 1
Dieser Artikel
220142202422029
description
SWAGELOK® 400-S, brass, 1/4 in. SWAGELOK
description
SWAGELOK® 200-S, brass, 1/8 in. SWAGELOK
description
SWAGELOK® 100-S, brass, 1/16 in. SWAGELOK
description
SWAGELOK® 400-S, stainless steel, 1/4 in. SWAGELOK
fitting
1/4 in. SWAGELOK
fitting
1/8 in. SWAGELOK
fitting
1/16 in. SWAGELOK
fitting
1/4 in. SWAGELOK
packaging
pkg of 10 ea
packaging
pkg of 10 ea
packaging
pkg of 10 ea
packaging
pkg of 5 ea
technique(s)
gas chromatography (GC): suitable using :Suitable
technique(s)
gas chromatography (GC): suitable using :Suitable
technique(s)
-
technique(s)
gas chromatography (GC): suitable using :Suitable
manufacturer/tradename
SWAGELOK® 400-S
manufacturer/tradename
SWAGELOK® 200-S
manufacturer/tradename
SWAGELOK® 100-S
manufacturer/tradename
SWAGELOK® 400-S
material
brass
material
brass
material
brass
material
stainless steel
Still not finding the right product?
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)