Fortfahren mit
Z763136
Prism centrifuge adapters
for 0.4 mL tubes, pack of 8 ea
Synonym(e):
lab equipment accessory
Anmelden zur Ansicht der Organisations- und Vertragspreise.
Über diesen Artikel
NACRES:
NB.22
UNSPSC Code:
41103911
Compatibility:
for use with C1801, for use with Labnet Prism (TM) Mini Centrifuge
Manufacturer/tradename:
Labnet C1206-8
feature
Holds 0.4 mL tubes
packaging
pack of 8 ea
manufacturer/tradename
Labnet C1206-8
technique(s)
centrifugation: suitable
compatibility
for use with C1801, for use with Labnet Prism (TM) Mini Centrifuge
Legal Information
Prism is a trademark of Labnet Intl., Inc.
Still not finding the right product?
Explore all of our products under Prism centrifuge adapters
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)