Merck
Z759848
KDS stainless steel syringes
volume 8 mL, with 1/16 in. Swagelok®
Synonym(e):
stainless steel syringes
Größe auswählen
1 EA
€ 1.130,00
€ 1.130,00
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
€ 1.130,00
About This Item
UNSPSC-Code:
41121500
NACRES:
NB.01
€ 1.130,00
Check Cart for Availability
Verpackung
pack of 1 ea
Hersteller/Markenname
KD Scientific 780802
Volumen
8 mL
Allgemeine Beschreibung
These rugged #316 stainless steel syringes are a perfect complement to the KDS 410 high pressure syringe dispenser. They eliminate the problem of breaking glass syringes when using high pressures. Compatibility with most chemicals is assured as wetted parts are #316 stainless steel, Viton® for the 20, 50 and 100 mL sizes, and Perfluoroelastomer for the 8 mL size.
Leistungsmerkmale und Vorteile
  • Compatible with most syringe pumps
  • Fully autoclavable
  • Good resistance to most chemicals
  • Safe solution when using high pressure dispensing
Rechtliche Hinweise
Swagelok is a registered trademark of Swagelok Company
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)