Fortfahren mit
Spend € 450 — Save 5 %; Spend € 700 — Save 10 %; Spend € 1.500 — Save 15 %*
Anmelden zur Überprüfung der Spielberechtigung. Produkte kaufen
Größe auswählen
| Größe/SKU | Verfügbarkeit | Preis |
|---|---|---|
1 ea | Warenkorb auf Verfügbarkeit prüfen | € 866,00 |
Über diesen Artikel
€ 866,00
Sparen Sie bis zu 15 %
feature
8-channel, electronic: no
packaging
pack of 1 ea
manufacturer/tradename
Sartorius 728120
volume
0.5-10 μL
Features and Benefits
- The click-stop system enables easy volume adjustment.
- Broad volume display for better reading.
- Color-coded for easy matching of fitting tips
- High chemical and UV-resistance materials to ensure a long lifetime of the pipette.
- Fully autoclavable and requires no disassembly.
- Easy cleaning and maintenance with only three parts to disassemble.
- Safe-cone filters provided for models >10μL.
- Εrgοnomic finger support reduces the grip force required to hold the pipette.
- Range of volume-3μL (5μL for fixed) to 10mL
- Low pipetting forces
- Wide range of adjustable single and multi-channel models and fixed single-channel models
Legal Information
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| description 8-channel, volume 0.5-10 μL | description 8-channel, volume 10-100 μL | description 8-channel, volume 30-300 μL | description 12-channel, volume 0.5-10 μL |
| feature electronic: no, 8-channel | feature electronic: no, 8-channel | feature electronic: no, 8-channel | feature electronic: no, 12-channel |
| manufacturer/tradename Sartorius 728120 | manufacturer/tradename Sartorius 728130 | manufacturer/tradename Sartorius 728140 | manufacturer/tradename Sartorius 728220 |
| volume 0.5-10 μL | volume 10-100 μL | volume 30-300 μL | volume 0.5-10 μL |
| packaging pack of 1 ea | packaging pack of 1 ea | packaging pack of 1 ea | packaging pack of 1 ea |
Still not finding the right product?
Explore all of our products under Sartorius Proline® Plus multichannel mechanical pippettor
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?