Merck
Z756954
Inhalation narcosis chamber
clear acrylic, pack of 1 ea
Größe auswählen
1 EA
€ 475,00
€ 475,00
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
€ 475,00
About This Item
UNSPSC-Code:
41100000
NACRES:
NB.22
€ 475,00
Check Cart for Availability
Materialien
clear acrylic
Verpackung
pack of 1 ea
Hersteller/Markenname
Bel-Art H46420-0002
Allgemeine Beschreibung
The Inhalation narcosis chamber has a space at the bottom filled with cotton soaked by anesthetic, and a vented floor directly above it to support the animal. Dimensions: I.D. is 197 mm (7 3/4 in.), top I.D. is 178 mm (7 in.), internal height is 127mm (5 in.), height below platform is 57 mm (2 1/4 in.), overall height is 197 mm (7 3/4 in.).
  • Optimally sized anesthetizing chamber for small animals including rats, mice, hamsters and guinea pigs
  • Body constructed of polypropylene and is unaffected by anesthesia chemicals
  • Features funnel for addition of anesthetic
  • Side-mounted stopcock lets fresh air enter to allow animal to breathe
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)