Ansicht ändern
| Größe/SKU | Verfügbarkeit | Preis |
|---|---|---|
100 ea | Warenkorb auf Verfügbarkeit prüfen | € 76,60 |
Über diesen Artikel
NACRES:
NB.04
UNSPSC Code:
41122410
eCl@ss:
32120613
Manufacturer/tradename:
Excel Scientific GP-PP-100
Material:
polypropylene
Sterility:
non-sterile
Packaging:
case of 100 ea
€ 76,60
Warenkorb auf Verfügbarkeit prüfen
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhaltenmaterial
polypropylene
sterility
non-sterile
feature
RNase and DNase free
packaging
case of 100 ea
manufacturer/tradename
Excel Scientific GP-PP-100
General description
For a more secure seal to minimize evaporation. General Purpose polypropylene films offer superior barrier properties that minimizes evaporation and prevents well-to-well contamination.
Application
Can be used to seal multiplates in general laboratory use, short-term storage, incubation and ELISA. Ideal for use with polypropylene plates.
Features and Benefits
- Certified DNase- & RNase- free
- Features good chemical resistance
- Provides a very secure seal to minimize evaporation, but is slightly harder to remove than SealPlate films
- Less rigid polypropylene film reduces aerosols upon removal
- Specially formulated adhesive minimizes plate residue
- Can be used as a temporary seal while awaiting further processing
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| description polypropylene, non-sterile | description polypropylene, sterile | description polyester | description polyester, W × L 3.125 in. × 5.750 in. |
| manufacturer/tradename Excel Scientific GP-PP-100 | manufacturer/tradename Excel Scientific GPS-PP-100 | manufacturer/tradename Excel Scientific SP-VS-100 | manufacturer/tradename Excel Scientific TTS-100 |
| sterility non-sterile | sterility sterile | sterility non-sterile | sterility non-sterile |
| material polypropylene | material polypropylene | material polyester , clear | material polyester |
| packaging case of 100 ea | packaging case of 100 ea | packaging pkg of 1 pack of 100 films | packaging pkg of 100 ea |
| feature RNase and DNase free | feature RNase and DNase free | feature - | feature - |
Still not finding the right product?
Explore all of our products under General Purpose Sealing Film
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?