Fortfahren mit
Z744067
FoilSeal™ Extra Thick Sealing Foil
aluminum, non-sterile
Anmelden zur Ansicht der Organisations- und Vertragspreise.
Größe auswählen
Ansicht ändern
Größe/SKUVerfügbarkeitPreis
100 ea
Warenkorb auf Verfügbarkeit prüfen
€ 109,00
Über diesen Artikel
NACRES:
NB.04
UNSPSC Code:
41122410
eCl@ss:
32120613
Manufacturer/tradename:
Excel Scientific F2-100
Material:
aluminum
W × l:
78 mm × 128 mm
Sterility:
non-sterile
Packaging:
case of 100 ea
€ 109,00
Warenkorb auf Verfügbarkeit prüfen
​
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten
material
aluminum
sterility
non-sterile
feature
RNase and DNase free
packaging
case of 100 ea
manufacturer/tradename
Excel Scientific F2-100
parameter
-40 °C temp. range (-20 °C)
W × L
78 mm × 128 mm
General description
Extra thick aluminum foil minimizes shredding upon removal. FoilSeal features a specially formulated adhesive that provides an excellent seal to minimize evaporation, reduce cross-contamination and prevent spillage. Universal fit for all plate types (e.g. flat-top, raised-rim and non-skirted); the smaller dimensions and rounded corners result in no plate overhang for robotics applications.
Helpful Tip: Remove diagonally from a corner to prevent tearing
FoilSeal sealing foils remove cleanly from plate surfaces with no adhesive stringing, residue or contamination. Each sealing foil measures 78.3 x 128 mm and offers sufficient sealing area for all plate types (e.g. flat top and raised-rim). Chamfered corners result in no plate overhang for robotics applications. Features one end-tab with perforations. Length between the perforation with end tab removed and the end of the foil seal is 118.1 mm.
  • Universal fit for use with all plate types (e.g. flat top and raised-rim)
  • The smaller dimensions and chamfered corners result in no plate overhang for robotics applications
  • Single end-tab features a custom perforation that will not tear during removal
  • Certified DNase- and RNase- free
  • Excellent barrier properties minimizes sample evaporation
  • Conformable foil allows visual verification of seal security
  • Pierceable with pipet tips and robotic probes for direct sample retrieval
Application
Multiplate sealing for PCR, light sensitive assays and robotics
Features and Benefits
  • Certified DNase- & RNase- free
  • Pierceable with a pipet tip or robotic probe for direct sample recovery
  • 2.0mil aluminum foil removes easily without shredding or tearing
  • The conformable nature of foil allows for the visual verification of the seal’s security around each well
  • Not recommended for long-term storage
Legal Information
FoilSeal is a trademark of Excel Scientific, Inc.
Ähnliche Artikel vergleichen
Vollständigen Vergleich anzeigen
Unterschiede anzeigen
1 of 1
Dieser Artikel
Z744068Z721549Z744071
description
aluminum, non-sterile
description
aluminum, sterile
description
size 38 μm, thick aluminum foil sealing film for use with 96 well plates, non-sterile
description
polypropylene, sterile
manufacturer/tradename
Excel Scientific F2-100
manufacturer/tradename
Excel Scientific F2S-50
manufacturer/tradename
Excel Scientific F-96-100
manufacturer/tradename
Excel Scientific GPS-PP-100
sterility
non-sterile
sterility
sterile
sterility
non-sterile
sterility
sterile
W × L
78 mm × 128 mm
W × L
78 mm × 128 mm
W × L
7.8 cm × 12.7 cm
W × L
-
material
aluminum
material
aluminum
material
aluminum film (medical grade adhesive)
material
polypropylene
packaging
case of 100 ea
packaging
case of 50 ea
packaging
pack of 100 ea
packaging
case of 100 ea
parameter
-40 °C temp. range (-20 °C)
parameter
-40 °C temp. range (120 °C)
parameter
-
parameter
-40-120 °C temp. range
Still not finding the right product?
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)