Ansicht ändern
| Größe/SKU | Verfügbarkeit | Preis |
|---|---|---|
36 ea | Warenkorb auf Verfügbarkeit prüfen | € 23,90 |
Über diesen Artikel
NACRES:
NB.35
UNSPSC Code:
55121611
Manufacturer/tradename:
Diversified Biotech PSTK-2030
Packaging:
pack of 36 ea
€ 23,90
Warenkorb auf Verfügbarkeit prüfen
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhaltenpackaging
pack of 36 ea
manufacturer/tradename
Diversified Biotech PSTK-2030
General description
Quick Reference - Petristickers correspond to wells on a microplate for fast colony identification
High Contrast - Colored grid allows colonies to stand out from the Petri dish background
Updated Design - Rectangular shape leaves ample space above and below grid for writing on the dish
Two Layouts - Reference wells numerically (1-96) or alpha numerically (A1-H12)
Ideally Sized - PetriStickers™ are 2.8 in. x 1.75 in. (71 mm x 44.5 mm), to fit most common Petri dishes
High Contrast - Colored grid allows colonies to stand out from the Petri dish background
Updated Design - Rectangular shape leaves ample space above and below grid for writing on the dish
Two Layouts - Reference wells numerically (1-96) or alpha numerically (A1-H12)
Ideally Sized - PetriStickers™ are 2.8 in. x 1.75 in. (71 mm x 44.5 mm), to fit most common Petri dishes
Legal Information
PetriStickers is a trademark of Diversified Biotech
p3xFLAG is a trademark of Sigma-Aldrich Co. LLC
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| description 16-sector pie format | description 12-sector pie format | description 6-sector pie format | description 32-square grid format |
| manufacturer/tradename Diversified Biotech PSTK-2030 | manufacturer/tradename Diversified Biotech PSTK-2020 | manufacturer/tradename Diversified Biotech PSTK-2000 | manufacturer/tradename Diversified Biotech PSTK-1000 |
| packaging pack of 36 ea | packaging pack of 36 ea | packaging pack of 36 ea | packaging pack of 36 ea |
Still not finding the right product?
Explore all of our products under PetriStickers™
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?