Ansicht ändern
| Größe/SKU | Verfügbarkeit | Preis |
|---|---|---|
25 ea | Warenkorb auf Verfügbarkeit prüfen | € 57,10 |
Über diesen Artikel
NACRES:
MA.01
UNSPSC Code:
41121800
Packaging:
pack of 25
Manufacturer/tradename:
Diversified Biotech MIWE-1000
€ 57,10
Warenkorb auf Verfügbarkeit prüfen
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhaltenpackaging
pack of 25
manufacturer/tradename
Diversified Biotech MIWE-1000
technique(s)
cell culture: suitable
well diam. × depth
1 cm × 0.25 mm
General description
Pre-cut, adhesive well stickers that firmly adhere to microscope slides. Examine specimens with or without cover slips.
Features and Benefits
- Convenient - Pre-cut, firmly adhere to microscope slides and enable specimen examination with or without cover slips.
- Multiple Applications - Examine or count tissue culture cells, yeast, bacteria and other microorganisms.
- Versatile - Compatible with different styles of microscopes (conventional, inverted or dissecting).
- Chemically Inert - Made of polyester with an acrylic adhesive. Microscope Slide-Wells are inert and stable under most laboratory conditions.
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| description well diam. × depth 1 cm × 0.25 mm, pack of 25 | description well diam. × depth 1 cm × 0.50 mm, pack of 25 | description use with dry or suspended specimens | description cut edges |
| manufacturer/tradename Diversified Biotech MIWE-1000 | manufacturer/tradename Diversified Biotech MIWE-2000 | manufacturer/tradename Diversified Biotech GRID-1000 | manufacturer/tradename BRAND 474701 |
| packaging pack of 25 | packaging pack of 25 | packaging pack of 40 ea | packaging - |
| well diam. × depth 1 cm × 0.25 mm | well diam. × depth 1 cm × 0.50 mm | well diam. × depth - | well diam. × depth - |
Still not finding the right product?
Explore all of our products under Microscope slide wells, 2-wells
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?