Größe auswählen
| Größe/SKU | Verfügbarkeit | Preis |
|---|---|---|
1 ea | Warenkorb auf Verfügbarkeit prüfen | € 162,00 |
Über diesen Artikel
€ 162,00
sterility
sterile; γ-irradiated
feature
Single chamber cell measuring 7.7cm3; Full assembly includes all necessary tubing.
manufacturer/tradename
IBI CFCAS0001
General description
The detachable tops also allow the researcher to coat the glass or plastic cover slip top with a variety of materials: such as seeded cells, proteins, polymer films, and/or ECM for experiments testing the response of biofilms or cells to different materials. As with other IBI flow cells, the convertible flow cell allows the user to monitor the biofilm in real time via standard or confocal laser microscopy. Complete convertible flow cell assemblies and single cells are available, and come gamma irradiated. The devices are single use only and cannot be autoclaved for re-use.
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| description Complete Assembly, Glass Cover Slip, sterile; γ-irradiated | description Complete Assembly, Plastic Cover Slip, sterile; γ-irradiated | description Cell Only, Glass Cover Slip, sterile; γ-irradiated | description Cell Only, Plastic Cover Slip, sterile; γ-irradiated |
| sterility sterile; γ-irradiated | sterility sterile; γ-irradiated | sterility sterile; γ-irradiated | sterility sterile; γ-irradiated |
| feature Single chamber cell measuring 7.7cm3; Full assembly includes all necessary tubing. | feature Single chamber cell measuring 7.7cm3; Full assembly includes all necessary tubing. | feature Flow Cell ONLY. No tubing included. | feature Flow Cell ONLY. No tubing included. |
| manufacturer/tradename IBI CFCAS0001 | manufacturer/tradename IBI CFCAS0002 | manufacturer/tradename IBI CFCAS0003 | manufacturer/tradename IBI CFCAS0004 |
Still not finding the right product?
Explore all of our products under Convertible flow cell
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?