Ansicht ändern
| Größe/SKU | Verfügbarkeit | Preis |
|---|
Über diesen Artikel
NACRES:
NB.35
UNSPSC Code:
46181503
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhaltensize
L
manufacturer/tradename
Arco 1763105
General description
MICROGARD™ 1500 PLUS white coverall is made from a highly breathable anti-static SMS fabric which utilises the latest developments in micro-fibre technology to ensure good filtration efficiency, offers protection from particles, perfect for working with asbestos, powders and general direct and dust with added light liquid spray protection.
Approved to:
EN ISO 13982-1 Type 5 Particulate protection
EN13034 Type 6 Reduced Spray
EN1149-5 Anti-static
EN1073-2 Barrier to radioactive particulates - Class 1
Approved to:
EN ISO 13982-1 Type 5 Particulate protection
EN13034 Type 6 Reduced Spray
EN1149-5 Anti-static
EN1073-2 Barrier to radioactive particulates - Class 1
Features and Benefits
- 3 piece hood
- 2-way front zipper with re-sealable storm flap
- Anti-static treatment
- Elasticated hood, wrists, waist and ankles
- Stitched seams
Legal Information
Microgard is a trademark of Microgard Limited
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| description size L | description size M | description size XL (elasticated ankles) | description size 2XL (boot ends with anti-slip soles) |
| size L | size M | size XL (elasticated ankles) | size 2XL (boot ends with anti-slip soles) |
| manufacturer/tradename Arco 1763105 | manufacturer/tradename Arco 1763104 | manufacturer/tradename Arco 1762006 | manufacturer/tradename Arco 1762307 |
Still not finding the right product?
Explore all of our products under MICROGARD™ 1500 PLUS coveralls for protection from particules and light spray
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?