Fortfahren mit
Z740130
BRAND® TipBox, empty
with blue tip-tray for 1000 μL tips
Synonym(e):
boxes for micropipette tips, empty boxes for pipette tips, empty tip box, pipette tip box
Anmelden zur Ansicht der Organisations- und Vertragspreise.
Größe auswählen
Ansicht ändern
Größe/SKUVerfügbarkeitPreis
1 ea
Warenkorb auf Verfügbarkeit prüfen
€ 8,00
Über diesen Artikel
NACRES:
NB.01
UNSPSC Code:
41122800
Manufacturer/tradename:
BRAND 732996
Feature:
autoclavable (121°C, 2 bar), Holds <1000 μl tips, stackable
Material:
colorless polypropylene box, lid, and holder (includes a colored tip-tray)
Packaging:
box of 1
€ 8,00
Warenkorb auf Verfügbarkeit prüfen
​
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten
material
colorless polypropylene box, lid, and holder (includes a colored tip-tray)
Quality Segment
sterility
non-sterile
feature
autoclavable (121°C, 2 bar), Holds <1000 μl tips, stackable
packaging
box of 1
manufacturer/tradename
BRAND 732996
General description
TipBox, non-sterile: the 5 mL and 10 mL tips are only available as a racked tip version in these specially corresponding TipBoxes.
Other Notes
BR732996 (India)
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Ähnliche Artikel vergleichen
Vollständigen Vergleich anzeigen
Unterschiede anzeigen
1 of 1
Dieser Artikel
Z740133Z740132Z740131
description
with blue tip-tray for 1000 μL tips
description
with yellow tip-tray for 200 μL tips
description
with green tip-tray for 300 μL tips
description
with gray tip-tray for pipette and filter tips up to 50 μL
manufacturer/tradename
BRAND 732996
manufacturer/tradename
BRAND 732992
manufacturer/tradename
BRAND 732994
manufacturer/tradename
BRAND 732990
feature
autoclavable (121°C, 2 bar), Holds <1000 μl tips, stackable
feature
Holds <200 μl tips, autoclavable (121°C, 2 bar), stackable
feature
autoclavable (121°C, 2 bar), Holds <300 μl tips, stackable
feature
stackable, autoclavable (121°C, 2 bar), Holds <50 μl tips
material
colorless polypropylene box, lid, and holder (includes a colored tip-tray)
material
colorless polypropylene box, lid, and holder (includes a colored tip-tray)
material
colorless polypropylene box, lid, and holder (includes a colored tip-tray)
material
colorless polypropylene box, lid, and holder (includes a colored tip-tray)
packaging
box of 1
packaging
box of 1
packaging
box of 1
packaging
box of 1
sterility
non-sterile
sterility
non-sterile
sterility
non-sterile
sterility
non-sterile
Quality Level
200
Quality Level
200
Quality Level
-
Quality Level
200
Still not finding the right product?
Explore all of our products under BRAND® TipBox, empty
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)