Größe auswählen
| Größe/SKU | Verfügbarkeit | Preis |
|---|---|---|
12 ea | Warenkorb auf Verfügbarkeit prüfen | € 356,00 |
Über diesen Artikel
€ 356,00
material
polycarbonate
feature
autoclavable, polycarbonate rotor-stator
packaging
case of 12, pack of 12
manufacturer/tradename
IKA 3452200
diam.
10 mm
Other Notes
Legal Information
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| description dispersing element for T-25 digital disperser, 10 mm, 10/cs | description dispersing element for T-25 digital disperser, 25 mm, 1/cs | description dispersing element for T-18 basic disperser, 10 mm, 1/cs | description dispersing element for T-10 basic disperser, 5 mm, 1/cs |
| manufacturer/tradename IKA 3452200 | manufacturer/tradename IKA 0001713800 | manufacturer/tradename IKA L004639 | manufacturer/tradename IKA 3304000 |
| material polycarbonate | material stainless steel | material stainless steel | material stainless steel |
| diam. 10 mm | diam. 25 mm | diam. 10 mm | diam. 5 mm |
| packaging case of 12, pack of 12 | packaging pack of 1 | packaging case of 1, pack of 1 | packaging case of 1, pack of 1 |
| feature polycarbonate rotor-stator | feature stainless steel rotor/stator | feature stainless steel rotor/stator | feature stainless steel rotor/stator |
Still not finding the right product?
Explore all of our products under IKA® ULTRA-TURRAX® disperser tools, plastic
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?